ContactUsLinkComponent
Hardness reagent, UniVer® 3
  HACH21320H
 :  Hach
 :  
undefined
Hardness reagent, UniVer® 3
  HACH21320H
 :  Hach
 :  
  • Chemical method:
    EDTA
  • Pk:
    28,3 g
  • Description:
    Hardness reagent, UniVer® 3
This combined indicator and buffer reagent is used for determination of total hardness by titration methods.