ContactUsLinkComponent
Nitrite kits, TNT and TNT plus kit
Nitrite kits, TNT and TNT plus kit
Catalog # 705-0163
Supplier:  Hach
CAS Number:  
undefined
Nitrite kits, TNT and TNT plus kit
Catalog # 705-0163
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    Diazotisation
  • Pk:
    1 KIT
  • Description:
    NitriVer 3 TNT Reagent Set
  • Seq:
    02
  • For unit:
    DR 900
  • Range:
    0,003 - 0,500 mg/l
Specifications
About this item
These 'Test N Tube' kits are used for the determination of nitrite by the diazotisation method.
  • Test 'N Tube products provide safe, convenient testing
  • TNT plus vials reduce errors with bar coded vials and RFID
  • TNT plus vials require no reagent blank