Order Entry
Czech Republic
ContactUsLinkComponent
Vertical electrophoresis system, omniPAGE Maxi
Vertical electrophoresis system, omniPAGE Maxi
Catalog # CLEVVS20DSYS
Supplier:  CLEAVER SCIENTIFIC
Vertical electrophoresis system, omniPAGE Maxi
Catalog # CLEVVS20DSYS
Supplier:  CLEAVER SCIENTIFIC

Specifications

  • Description:
    Vertical electrophoresis system, omniPAGE maxi, with casting base
  • Model:
    omniPAGE maxi
  • Glass plate size (W×H):
    200×200 mm
  • Gel size W×H:
    160×175 mm
  • Unit W×H×D:
    260×280×160 mm
  • Buffer volume, gel tank:
    1200-5600 ml
  • Pk:
    1

Specifications

About this item

omniPAGE maxi combines convenient ease of use features with high resolution separations. Simple set up using ultra soft silicone seals guarantees trouble free glass plate loading and gel casting. Dual gaskets on the gel running insert along with notched and plain glass plates ensure leak proof gel running. Rapid set up cooling retains resolution in extended separations and also saves on buffer volume without affecting run quality. Glass plates reduce breakage and have bonded spacers for added convenience.

  • Ideal for high resolution separations
  • Rapid gel casting and loading
  • Optional low or high buffer volumes
  • Rapid set up cooling
Delivery: Each complete system includes glass plates with bonded 1,0 mm thick spacers, 2× 24 sample combs, cooling pack and dummy plate.