Barium sulphate USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:7727-43-7
:
Formula:BaSO₄
MDL Number:MFCD00003455
Boiling Pt:1600
Melting Pt:1345
Density:4.5 g/cm³ (25 °C)
Molecular Weight:233.39 g/mol
Merck Index:13,00997
Storage Temperature:Ambient
EINECS:231-784-4
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:97.5% Min.
Specification Test Results
Frequently Bought Together