Order Entry
Czech Republic
ContactUsLinkComponent
 
Amantadine hydrochloride USP
Catalog # SPCMAM102-1KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Amantadine hydrochloride USP
Catalog # SPCMAM102-1KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specification Test Results

About this item

 :
 :187.71 g/mol
Melting Pt:360 °C
EINECS:211-560-2
CAS Number:665-66-7
Formula:C₁₀H₁₈ClN
MDL Number:MFCD00074723