
Docusate sodium USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:577-11-7
:
Formula:C₂₀H₃₇NaO₇S
MDL Number:MFCD00012455
:444.57 g/mol
Merck Index:12,03460
Boiling Pt:∼180 °C (1013 hPa)
Melting Pt:∼155 °C
Density:∼1.1 g/cm³ (20 °C)
EINECS:209-406-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:99% Min.
Specification Test Results
Frequently Bought Together