N-Boc-1,6-hexanediamine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:65915-94-8
:
Formula:C₁₁H₂₄N₂O₂·HCl
MDL Number:MFCD00039072
:252.783 g/mol
Melting Pt:∼ 164 °C (dec.)
Storage Temperature:Ambient
EINECS:265-976-4
Related Information
Frequently Bought Together