
7-Methylindole-3-carboxyaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4771-50-0
:
Formula:C₁₀H₉NO
MDL Number:MFCD00047170
:159.19 g/mol
Melting Pt:209…211 °C
:GHS07
Related Information
Frequently Bought Together