Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:72022-68-5
:
Formula:C₂₂H₄₀N₂S₁₀Zn
MDL Number:MFCD00623815
:718.60 g/mol
Melting Pt:202…206 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together