
Ultramark® 1621 mass spec standard
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105809-15-2
:
Formula:C₆H₁₈N₃P₃
MDL Number:MFCD00242454
:231.189 g/mol
Density:1.8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together