
Diethyl tetrafluorosuccinate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:377-71-9
:
Formula:C₈H₁₀F₄O₄
MDL Number:MFCD00015155
:246.16 g/mol
Boiling Pt:78 °C (5 mmHg)
Density:1.273
EINECS:206-821-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together