


Methylenediphenyl-4,4'-diisocyanate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:101-68-8
:
Formula:C₁₅H₁₀N₂O₂
MDL Number:MFCD00036131
:250.26 g/mol
Boiling Pt:194 °C (5 mmHg)
Melting Pt:41…44 °C
Density:1.239
Flash Pt:196 °C (385 °F)
Storage Temperature:Freezer
EINECS:202-966-0
UN:2811
ADR:6.1,II
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together