
3-Fluoro-DL-valine 94%
Katalogové číslo L15334.MC
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:135.14 g/mol
Melting Pt:∼ 250 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:43163-94-6
Formula:C₅H₁₀FNO₂
MDL Number:MFCD00077549