

Cyclopropanecarboxaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1489-69-6
:
Formula:C₄H₆O
MDL Number:MFCD00012261
:70.09 g/mol
Boiling Pt:98…101 °C
Density:0.938
Flash Pt:7 °C (44 °F)
Storage Temperature:Ambient
ADR:3,II
:GHS02
:GHS05
Related Information
Frequently Bought Together