

Cyclohexanemethyl isothiocyanate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52395-66-1
:
Formula:C₈H₁₃NS
MDL Number:MFCD00040881
:155.26 g/mol
Boiling Pt:222…223 °C
Density:1.020
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together