
DL-2,3-Diaminopropionic acid hydrochloride 99%
Catalog # L09485.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:140.57 g/mol
Merck Index:14,2984
Melting Pt:∼ 232 °C (dec.)
EINECS:259-387-1
:GHS07
Storage Temperature:Ambient
CAS Number:54897-59-5
Formula:C₃H₈N₂O₂
MDL Number:MFCD00012884