Diethyl isobutylmalonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10203-58-4
:
Formula:C₁₁H₂₀O₄
MDL Number:MFCD00026869
:216.28 g/mol
Boiling Pt:225 °C
Density:0.970
Flash Pt:>100 °C (212 °F)
Storage Temperature:Ambient
EINECS:233-504-6
Related Information
Frequently Bought Together