1,2-Diphenylcyclopropane cis- and trans-mixture 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29881-14-9
:
Formula:C₁₅H₁₄
MDL Number:MFCD00040875
:194.28 g/mol
Boiling Pt:175…177 °C (14 mmHg)
Density:1.020
Storage Temperature:Ambient
Related Information
Frequently Bought Together