
Tetra-n-dodecylammonium iodide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40797-39-5
:
Formula:C₄₈H₁₀₀IN
MDL Number:MFCD00051856
:818.23 g/mol
Melting Pt:114…118 °C
EINECS:255-086-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together