
trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54125-02-9
:
Formula:C₈H₁₆O₂Si
MDL Number:MFCD00008498
Merck Index:14,6001
Boiling Pt:68…69 °C (14 mmHg)
Density:0.890
Flash Pt:45 °C (113 °F)
ADR:3,III
:172.299 g/mol
:GHS02
Related Information
Frequently Bought Together