Diethyl-1-naphthylmethylphosphonate ≥90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:53575-08-9
:
Formula:C₁₅H₁₉O₃P
MDL Number:MFCD00015769
Boiling Pt:205…206 °C (5 mmHg)
:278.29 g/mol
Storage Temperature:Ambient
Frequently Bought Together