
Methyl adipoyl chloride 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35444-44-1
:
Formula:C₇H₁₁ClO₃
MDL Number:MFCD00013661
Boiling Pt:76 °C (0.8 mmHg)
Density:1.050
EINECS:252-569-1
ADR:8,II
:178.62 g/mol
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together