
4,4'-cis-Stilbenedicarboxylic acid 95%
Catalog # L04364.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:>300 °C
:268.27 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:133005-88-6
Formula:C₁₆H₁₂O₄
MDL Number:MFCD00013994