
exo-3,6-Epoxy-1,2,3,6-tetrahydrophthalic anhydride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6118-51-0
:
Formula:C₈H₆O₄
MDL Number:MFCD00151506
:166.13 g/mol
Melting Pt:118…119 °C
EINECS:226-570-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together