S,S-Dimethyl-N-(p-toluenesulphonyl)sulphoximine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22236-45-9
:
Formula:C₉H₁₃NO₃S₂
MDL Number:MFCD00013922
:247.34 g/mol
Melting Pt:167…169 °C
Storage Temperature:Ambient
EINECS:244-861-2
Related Information
Frequently Bought Together