Order Entry
Czech Republic
ContactUsLinkComponent
Methyl (methylsulphinyl)methyl sulphide 97%
Methyl (methylsulphinyl)methyl sulphide 97%
Catalog # L00510.09
CAS Number:  
Methyl (methylsulphinyl)methyl sulphide 97%
Catalog # L00510.09
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :124.22 g/mol
Boiling Pt:222…226 °C
Density:1.222
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:251-577-2
CAS Number:33577-16-1
Formula:C₃H₈OS₂
MDL Number:MFCD00002091