Gold 95% supplied in 0.1 mM PBS, nanoparticles 50 nm OD1 (Abs. 535 nm)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-57-5
:
Formula:Au
MDL Number:MFCD00003436
:196.97 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together