Gold 95% supplied in 0.1 mM PBS, nanoparticles 15 nm OD1 (Abs. 520 nm)
Katalogové číslo J67141.AC
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:196.97 g/mol
Storage Temperature:Refrigerator
CAS Number:7440-57-5
Formula:Au
MDL Number:MFCD00003436