
Meropenem trihydrate 98-101%
Catalog # J66446.MF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
Specifications
About this item
:
:437.51 g/mol
Merck Index:14,5900
Boiling Pt:627 °C
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Refrigerator
CAS Number:119478-56-7
Formula:C₁₇H₃₁N₃O₈S
MDL Number:MFCD08600005