Order Entry
Czech Republic
ContactUsLinkComponent
 
Toxic
Health
Cycloheximide 100 mg/ml in DMSO
Catalog # J66004.XF
CAS Number:  
undefined
Toxic
Health
Cycloheximide 100 mg/ml in DMSO
Catalog # J66004.XF
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :281.35 g/mol
ADR:6.1,II
 :Filtered through a 0.2 µm filter.
Flash Pt:87°0 °C (230 °F)
EINECS:200-636-0
UN:2810
 :GHS06
 :GHS08
Storage Temperature:Refrigerator
CAS Number:66-81-9
Formula:C₁₅H₂₃NO₄
MDL Number:MFCD00082346