Order Entry
Czech Republic
ContactUsLinkComponent
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine 98%
Toxic
Irritant
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine 98%
Catalog # J65960.03
CAS Number:  
Toxic
Irritant
5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine 98%
Catalog # J65960.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :530.57 g/mol
EINECS:245-814-9
 :GHS06
 :GHS07
CAS Number:23669-79-6
Formula:C₃₀H₃₀N₂O₇
MDL Number:MFCD00063410