Order Entry
Czech Republic
ContactUsLinkComponent
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine 98%
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine 98%
Catalog # J65402.03
CAS Number:  
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine 98%
Catalog # J65402.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Formula:C₃₈H₃₄FN₅O₆
MDL Number:MFCD15145191
 :675.70 g/mol
Storage Temperature:Refrigerator