Order Entry
Czech Republic
ContactUsLinkComponent
 
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine 98%
Catalog # J64880.03
CAS Number:  
5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine 98%
Catalog # J64880.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :560.59 g/mol
Storage Temperature:Refrigerator
Formula:C₃₁H₃₂N₂O₈
MDL Number:MFCD00274123