Order Entry
Czech Republic
ContactUsLinkComponent
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine 98%
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine 98%
Catalog # J64217.06
CAS Number:  
5'-O-(4,4'-Dimethoxytrityl)-2'-fluoro-2'-deoxyuridine 98%
Catalog # J64217.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :548.56 g/mol
CAS Number:146954-74-7
Formula:C₃₀H₂₉N₂O₇F
MDL Number:MFCD06657648