
N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite) 98%
Catalog # J64064.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:875.92 g/mol
:GHS07
CAS Number:136834-22-5
Formula:C₄₇H₅₁FN₇O₇P
MDL Number:MFCD12912392