
Proadifen hydrochloride
Catalog # J63833.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
CAS Number:62-68-0
:
Formula:C₂₃H₃₁NO₂·HCl
:389.964 g/mol
Melting Pt:122…123 °C
:GHS07
MDL Number:MFCD00055151