Order Entry
Czech Republic
ContactUsLinkComponent
Ambroxol hydrochloride 98%
Ambroxol hydrochloride 98%
Catalog # J63712.06
CAS Number:  
Ambroxol hydrochloride 98%
Catalog # J63712.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

 :
 :414.57 g/mol
Merck Index:14,386
EINECS:242-500-3
CAS Number:23828-92-4
Formula:C₁₃H₁₈Br₂N₂O.HCl
MDL Number:MFCD00078932