

Triamcinolone acetonide ≥98%
Catalog # J63548.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
Specifications
About this item
:
:434.504 g/mol
Merck Index:14,9596
Melting Pt:274…278 °C
EINECS:200-948-7
:GHS08
:GHS07
CAS Number:76-25-5
Formula:C₂₄H₃₁FO₆
MDL Number:MFCD00056834