Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bromhexine hydrochloride
Bromhexine hydrochloride ≥98%
Bromhexine hydrochloride ≥98%
:
Thermo Fisher Scientific
Bromhexine hydrochloride ≥98%
:
Thermo Fisher Scientific
:
611-75-6
:
:
C₁₄H₂₀Br₂N₂
:
MFCD00056626
:
412.59 g/mol
:
14,1391
:
240…244 °C
:
Ambient
:
210-280-8
msds_ALFAJ63451
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
n-Pentan ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
: Merck
Aceton ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
1-Propanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Pyridin, EMSURE® ACS, Reag. Ph. Eur. p.a., Supelco®
: Merck
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
: Merck
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
terc-Butylmethylether, EMSURE® ACS p.a., Supelco®
: Merck
Glycerin ≥98%, GPR RECTAPUR®
: VWR Chemicals
Dichlormethan ≥98% stabilised, TECHNICAL
: VWR Chemicals
Tetrahydrofuran ≥99.5% (by anhydrous basis) stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Ethylacetát ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethylacetát ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Diethylether stabilizovaný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
: Merck
Aceton, EMPARTA® ACS analytical reagent, Supelco®
: Merck