Order Entry
Czech Republic
ContactUsLinkComponent
Triton® X-100 lysis buffer, pH 7,4
Triton® X-100 lysis buffer, pH 7,4
Certificates
Triton® X-100 lysis buffer, pH 7,4
Certificates

Specifications

  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Related Information

Frequently Bought Together