Order Entry
Czech Republic
ContactUsLinkComponent
 
Sodium pyrophosphate, 200 mM buffer solution
 
Sodium pyrophosphate, 200 mM buffer solution
 

 

  • CAS No.:
    7758-16-9
  • MDL No.:
    MFCD00014246
  • Physical form:
    Liquid

 

Frequently Bought Together