
Isoproterenol hydrochloride 98%
Katalogové číslo J61788.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
Specifications
About this item
:
:247.72 g/mol
Merck Index:14,5217
Melting Pt:165…175 °C (dec.)
EINECS:200-089-8
:GHS07
Storage Temperature:Ambient
CAS Number:51-30-9
Formula:C₁₁H₁₇NO₃
MDL Number:MFCD00012603