Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nω-Methyl-L-arginine monoacetate
Nω-Methyl-L-arginine monoacetate ≥99%
Nω-Methyl-L-arginine monoacetate ≥99%
J60890.MF
:
Thermo Fisher Scientific
:
Nω-Methyl-L-arginine monoacetate ≥99%
J60890.MF
:
Thermo Fisher Scientific
:
Pk:
50 mg
:
:
248.28 g/mol
:
14,6022
:
Ambient
:
53308-83-1
:
C₇H₁₆N₄O₂·CH₃COOH
:
MFCD00069311