
(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride
Catalog # J60695.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
Specifications
About this item
:
:277.23 g/mol
Merck Index:14,3720
EINECS:213-970-7
:GHS08
Storage Temperature:Ambient
CAS Number:1070-11-7
Formula:C₁₀H₂₄N₂O₂·₂HCl
MDL Number:MFCD00216025