Order Entry
Czech Republic
ContactUsLinkComponent
Tacrine hydrochloride ≥98%
Toxic
Irritant
Tacrine hydrochloride ≥98%
Catalog # J60070.06
CAS Number:  
Toxic
Irritant
Tacrine hydrochloride ≥98%
Catalog # J60070.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :234.72 g/mol
Merck Index:14,9024
Melting Pt:283…284 °C
EINECS:216-867-5
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:1684-40-8
Formula:C₁₃H₁₄N₂·HCl
MDL Number:MFCD00012657