Order Entry
Czech Republic
ContactUsLinkComponent
3-Iodo-1-methyl-1H-pyrazole 97%
Corrosive
Irritant
3-Iodo-1-methyl-1H-pyrazole 97%
Katalogové číslo H66356.03
CAS Number:  
Corrosive
Irritant
3-Iodo-1-methyl-1H-pyrazole 97%
Katalogové číslo H66356.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

 :
 :208.00 g/mol
Density:1.936
Flash Pt:110 °C (230 °F)
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:92525-10-5
Formula:C₄H₅IN₂
MDL Number:MFCD08435890