2-(4-Bromophenyl)propan-2-ol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2077-19-2
:
Formula:C₉H₁₁BrO
MDL Number:MFCD11870096
:215.09 g/mol
Boiling Pt:75…85 °C (0,1 mmHg)
Melting Pt:40…45 °C
Density:1,356
Storage Temperature:Ambient
Related Information
Frequently Bought Together