N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123847-85-8
:
Formula:C₄₄H₃₂N₂
MDL Number:MFCD03093246
:588.75 g/mol
Melting Pt:279…283 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together