
2-Bromo-6-fluoro-4-methylpyridine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:180608-37-1
:
Formula:C₆H₅BrFN
MDL Number:MFCD09839269
:190.02 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together