Order Entry
Czech Republic
ContactUsLinkComponent
5-Bromo-2-chlorobenzaldehyde 98%
Irritant
5-Bromo-2-chlorobenzaldehyde 98%
Catalog # H64438.06
CAS Number:  
Irritant
5-Bromo-2-chlorobenzaldehyde 98%
Catalog # H64438.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

 :
 :219.46 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:189628-37-3
Formula:C₇H₄BrClO
MDL Number:MFCD08445659